2-Chloro-4-methylpyrimidine

Supplier: APOLLO SCIENTIFIC
Danger

OR12067-100G OR12067-25G OR12067-5G
77404-146EA 124.3 USD
77404-146 77404-148 77404-150
2-Chloro-4-methylpyrimidine
2-Chloro-4-methylpyrimidine
Formula: C₅H₅ClN₂
Boiling Pt: 93…94 °C/17 mmHg
Melting Pt: 45…53 °C
Flash Pt: 98 °C
Storage Temperature: Freezer
MDL Number: MFCD00054434
CAS Number: 13036-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR