Potassium thioacetate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Potassium ethanethioate

OR59966-100G OR59966-25G
77410-646EA 41.78 USD
77410-646 77410-648
Potassium thioacetate
Potassium thioacetate
Formula: C₂H₃KOS
MW: 114.21 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00137704
CAS Number: 10387-40-3
EINECS: 233-848-7
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR