3,4-(Methylenedioxy)aniline, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-1,3-benzodioxole

OR28575-100G OR28575-25G OR28575-5G
77408-094EA 69.28 USD
77408-094 77408-096 77408-098
3,4-(Methylenedioxy)aniline, Technical Grade
3,4-(Methylenedioxy)aniline
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 144 °C/16 mmHg
Melting Pt: 40…45 °C
Flash Pt: 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005832
CAS Number: 14268-66-7
EINECS: 238-161-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR