Methyl-4,6-dichloronicotinate 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 4,6-dichloronicotinate

OR14223-1G OR14223-25G OR14223-5G
77404-858EA 21.26 USD
77404-858 77404-860 77404-862
Methyl-4,6-dichloronicotinate 95+%
Methyl-4,6-dichloronicotinate
Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 41…43 °C
Storage Temperature: Freezer
MDL Number: MFCD04125732
CAS Number: 65973-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR