Ethyl-5-bromonicotinate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 5-bromonicotinate

OR5075-100G OR5075-500G
77410-124EA 103.92 USD
77410-124 77410-126
Ethyl-5-bromonicotinate 98%
Ethyl-5-bromonicotinate
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Boiling Pt: 86…92 °C/0.5 mmHg
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00040366
CAS Number: 20986-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR