1,3-Dibromo-5,5-dimethylhydantoin 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR10135-500G OR10135-100G
77402-494EA 66.22 USD
77402-494 77402-492
1,3-Dibromo-5,5-dimethylhydantoin 98%
1,3-Dibromo-5,5-dimethylhydantoin
Formula: C₅H₆Br₂N₂O₂
Melting Pt: 197…199 °C (dec.)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00003189
CAS Number: 77-48-5
EINECS: 201-030-9
UN: 3087
ADR: 5.1+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR