6-Chloro-2-pyridineboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Chloropyridine-2-boronic Acid

OR2262-1G OR2262-250MG
77406-916EA 241.4 USD
77406-916 77406-918
6-Chloro-2-pyridineboronic acid
6-Chloro-2-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063028
CAS Number: 652148-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR