(R)-(-)-β-Aminophenethyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

OR19518-100G OR19518-25G OR19518-5G
77406-474EA 143.66 USD
77406-474 77406-476 77406-478
(R)-(-)-β-Aminophenethyl alcohol 97%
(R)-(-)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00008062
CAS Number: 56613-80-0
EINECS: 260-287-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR