5-Fluoro-2-methoxypyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC10417-10G PC10417-1G PC10417-5G
77412-882EA 59.1 USD
77412-882 77412-884 77412-886
5-Fluoro-2-methoxypyridine 97%
5-Fluoro-2-methoxypyridine
Formula: C₆H₆FNO
Boiling Pt: 137…140 °C
Flash Pt: <60 °C
Storage Temperature: Freezer
MDL Number: MFCD03095252
CAS Number: 51173-04-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR