2-Fluoro-6-methoxypyridine

Supplier: APOLLO SCIENTIFIC
Warning

PC3181-25G PC3181-5G
77414-564EA 80.48 USD
77414-564 77414-566
2-Fluoro-6-methoxypyridine
2-Fluoro-6-methoxypyridine
Formula: C₆H₆FNO
Flash Pt: <60 °C
Storage Temperature: Ambient
MDL Number: MFCD09998161
CAS Number: 116241-61-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR