Triethyloxonium tetrafluoroborate 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC7110-100G PC7110-25G
77417-638EA 323.98 USD
77417-638 77417-640
Triethyloxonium tetrafluoroborate 95%
Triethyloxonium tetrafluoroborate

Powerful alkylating reagent for many reactions.

Formula: C₆H₁₅BF₄O
MW: 189.99 g/mol
Melting Pt: 84…89 °C
Density: 1.328 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044423
CAS Number: 368-39-8
EINECS: 206-705-1
UN: 2925
ADR: 4.1+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR