3-Chlor-1,2-benzisothiazol

Supplier: APOLLO SCIENTIFIC
Warning

OR2649-100G OR2649-25G OR2649-5G
77407-794EA 407.52 USD
77407-794 77407-796 77407-798
3-Chlor-1,2-benzisothiazol
3-Chlor-1,2-benzisothiazol
Formula: C₇H₄ClNS
Boiling Pt: 80…86 °C/0.75 mmHg
Melting Pt: 38…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00673254
CAS Number: 7716-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR