Bismuth(III) trifluoromethanesulphonate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Bismuth triflate, Bismuth(III) trifluoromethanesulphonate

PC9890-100G PC9890-25G PC9890-5G
77418-746EA 231.28 USD
77418-746 77418-748 77418-750
Bismuth(III) trifluoromethanesulphonate
Bismuth(III) trifluoromethanesulphonate
Formula: C₃BiF₉O₉S₃
MW: 656.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02093669
CAS Number: 88189-03-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR