Ytterbium(III) trifluoromethanesulphonate hydrate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Trifluoromethanesulphonic acid ytterbium salt hydrate, Ytterbium(III) trifluoromethanesulphonate hydrate, Ytterbium(III) triflate hydrate, Ytterbium triflate hydrate

PC49155-10G PC49155-25G
77415-872EA 84.56 USD
77415-872 77415-874
Ytterbium(III) trifluoromethanesulphonate hydrate 98%
Ytterbium(III) trifluoromethanesulphonate hydrate
Formula: C₃F₉O₉S₃Yb·xH₂O
MW: 620.25 g/mol (anhy)
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD03703499
CAS Number: 252976-51-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR