2-Furoic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Furan-2-carboxylic acid, 2-Furancarboxylic acid

OR10949-500G OR10949-100G OR10949-25G
77402-994EA 28.5 USD
77402-994 77402-990 77402-992
2-Furoic acid 95%
2-Furoic acid
Formula: C₅H₄O₃
MW: 112.08 g/mol
Boiling Pt: 230 °C
Melting Pt: 127…132 °C
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00003238
CAS Number: 88-14-2
EINECS: 201-803-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR