4-Bromosalicylic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Bromo-2-hydroxybenzoic acid

OR15002-100G OR15002-25G OR15002-5G
77405-224EA 156.9 USD
77405-224 77405-226 77405-228
4-Bromosalicylic acid 98%
4-Bromosalicylic acid
Formula: C₇H₅BrO₃
MW: 217.02 g/mol
Melting Pt: 216…217 °C
Storage Temperature: Ambient
MDL Number: MFCD06797970
CAS Number: 1666-28-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR