2,6-Difluoro-3-nitrobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2863-25G PC2863-5G
77413-924EA 41.78 USD
77413-924 77413-926
2,6-Difluoro-3-nitrobenzonitrile 98%
2,6-Difluoro-3-nitrobenzonitrile
Formula: C₇H₂F₂N₂O₂
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00042254
CAS Number: 143879-77-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR