3-Bromothiophene-2-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene

OR29108-1G OR29108-25G OR29108-5G
77408-328EA 21.26 USD
77408-328 77408-330 77408-332
3-Bromothiophene-2-carbaldehyde
3-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 110…113 °C/10 mmHg
Melting Pt: 24…25 °C
Density: 1.755 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00126680
CAS Number: 930-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR