(S)-(-)-4-Chloro-α-methylbenzylamine 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: (S)-1-(4-Chlorophenyl)ethylamine

OR911668-1G OR911668-5G
77411-620EA 75.4 USD
77411-620 77411-622
(S)-(-)-4-Chloro-α-methylbenzylamine 98%
(S)-(-)-4-Chloro-α-methylbenzylamine
Formula: C₈H₁₀ClN
MW: 155.63 g/mol
Boiling Pt: 232 °C/760 mmHg
Density: 1.115 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00671640
CAS Number: 4187-56-8
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR