2-Bromo-3,3,3-trifluoropropene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC1602-100G PC1602-25G PC1602-5G
77413-264EA 149.76 USD
77413-264 77413-266 77413-268
2-Bromo-3,3,3-trifluoropropene 98%
2-Bromo-3,3,3-trifluoropropene

Versatile CF3 building block: Hu et al, J. Chem Soc., Perkin Trans. 1, 1998, 2, 279. Precursor for PC77612-(Trifluoromethyl)acrylic acid : JOC., 1989 , 54.

Formula: C₃H₂BrF₃
Boiling Pt: 29…30 °C
Density: 1.66 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00077469
CAS Number: 1514-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR