1-Aminocyclopropanecarboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR0355T-100G OR0355T-25G OR0355T-5G
77402-056EA 475.78 USD
77402-056 77402-058 77402-060
1-Aminocyclopropanecarboxylic acid 97%
1-Aminocyclopropanecarboxylic acid

An NMDA receptor agonist acting at the glycine site.

Formula: C₄H₇NO₂
MW: 101.11 g/mol
Melting Pt: 229…231 °C
Storage Temperature: Freezer
MDL Number: MFCD00009944
CAS Number: 22059-21-8
EINECS: 606-917-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR