2-Hydroxyphenylboronic acid pinacol ester 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

OR11012-1G OR11012-5G
77403-090EA 21.26 USD
77403-090 77403-092
2-Hydroxyphenylboronic acid pinacol ester 98%
2-Hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
Boiling Pt: 282 °C
Density: 1.05 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093754
CAS Number: 269409-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR