2-Bromo-5-(trifluoromethyl)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2479-25G PC2479-5G
77413-656EA 85.58 USD
77413-656 77413-658
2-Bromo-5-(trifluoromethyl)benzonitrile 98%
2-Bromo-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
Boiling Pt: 107…115 °C/10 mmHg
Melting Pt: 50…51 °C
Storage Temperature: Ambient
MDL Number: MFCD03412187
CAS Number: 1483-55-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR