Ethyl indole-2-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester

OR10188-25G OR10188-5G
77402-536EA 23.36 USD
77402-536 77402-538
Ethyl indole-2-carboxylate 98%
Ethyl indole-2-carboxylate
Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 122…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005609
CAS Number: 3770-50-1
EINECS: 223-206-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR