6-Chloro-2-fluoro-m-cresol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 6-Chloro-2-fluoro-3-methylphenol

PC0119-100G PC0119-25G PC0119-5G
77412-394EA 1172.66 USD
77412-394 77412-396 77412-398
6-Chloro-2-fluoro-m-cresol 98%
6-Chloro-2-fluoro-m-cresol
Formula: C₇H₆ClFO
MW: 160.58 g/mol
Melting Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD01631376
CAS Number: 261762-91-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR