Isobutyryl chloride 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR10005-100G
OR10005-25G
OR10005-500G
77402-392EA
41.78
USD
77402-392
77402-394
77402-396
Isobutyryl chloride 98%
Isobutyryl chloride
Formula:
C₄H₇ClO Boiling Pt: 91…93 °C Melting Pt: –90 °C Density: 1.017 g/cm³ (25 °C) Flash Pt: 8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000717 CAS Number: 79-30-1 EINECS: 201-194-1 UN: 2395 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...