2,6-Dibromo-4-tert-butylaniline 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(tert-Butyl)-2,6-dibromoaniline, 2,6-Dibromo-4-(tert-butyl)aniline

OR1068-100G OR1068-25G OR1068-5G
77402-876EA 254.7 USD
77402-876 77402-878 77402-880
2,6-Dibromo-4-tert-butylaniline 97%
2,6-Dibromo-4-tert-butylaniline
Formula: C₁₀H₁₃Br₂N
MW: 307.03 g/mol
Boiling Pt: 136…140 °C/2 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD06657952
CAS Number: 10546-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR