Silver trifluoroacetate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Silver(I) trifluoroacetate

PC6440-100G PC6440-25G PC6440-5G
77417-386EA 144.68 USD
77417-386 77417-388 77417-390
Silver trifluoroacetate 98%
Silver trifluoroacetate

Lewis acid catalyst for regioselective alkylation:Helv.Chim.Acta., 1989, 72, 1362; Can convert enol silyl ethers to alpha-acyloxy carbonyls, ask for reference.

Formula: C₂AgF₃O₂
MW: 220.88 g/mol
Melting Pt: 257…260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013199
CAS Number: 2966-50-9
EINECS: 221-004-0
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR