2-Amino-4-chlorothiophenol 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Amino-4-chlorobenzenethiol, 2-Amino-4-chlorophenyl mercaptan, 5-Chloro-2-mercaptoaniline

OR0785-100G OR0785-25G OR0785-5G
77402-240EA 128.38 USD
77402-240 77402-242 77402-244
2-Amino-4-chlorothiophenol 97%
2-Amino-4-chlorothiophenol
Formula: C₆H₆ClNS
MW: 159.64 g/mol
Melting Pt: 56…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792528
CAS Number: 1004-00-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR