2,3-Difluoro-6-methoxyaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Amino-3,4-difluoroanisole, 5,6-Difluoro-o-anisidine

PC303015-1G PC303015-5G
77414-316EA 179.32 USD
77414-316 77414-318
2,3-Difluoro-6-methoxyaniline 98%
2,3-Difluoro-6-methoxyaniline
Formula: C₇H₇F₂NO
Storage Temperature: Ambient
CAS Number: 886501-51-5
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR