2,2,2-Trifluoroethyl acrylate 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
2,2,2-Trifluoroethylacrylate, Acrylic acid 2,2,2-trifluoroethyl ester
PC7316-100G
PC7316-25G
77417-806EA
62.16
USD
77417-806
77417-808
2,2,2-Trifluoroethyl acrylate 99%
2,2,2-Trifluoroethyl acrylate
Formula:
C₅H₅F₃O₂ MW: 154.09 g/mol Boiling Pt: 46 °C/125 mmHg Density: 1.216 g/cm³ (25 °C) Flash Pt: 12 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000444 CAS Number: 407-47-6 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...