2-Bromo-4-fluoro-5-(trifluoromethyl)aniline
Supplier: APOLLO SCIENTIFIC
Synonyms:
6-Bromo-α,α,α,4-tetrafluoro-m-toluidine, 5-Amino-4-bromo-2-fluorobenzotrifluoride
PC6197-10G
PC6197-1G
PC6197-5G
77417-306EA
189
USD
77417-306
77417-308
77417-310
2-Bromo-4-fluoro-5-(trifluoromethyl)aniline
2-Bromo-4-fluoro-5-(trifluoromethyl)aniline
Formula:
C₇H₄BrF₄N MW: 258.01 g/mol Melting Pt: 24…29 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973755 CAS Number: 193090-60-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...