Propiolaldehyde diethyl acetal 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne
OR54615-100G
OR54615-25G
OR54615-5G
77410-396EA
124.3
USD
77410-396
77410-398
77410-400
Propiolaldehyde diethyl acetal 95%
Propiolaldehyde diethyl acetal
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 38 Density: 0.894 g/cm³ (25 °C) Flash Pt: 31 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009237 CAS Number: 10160-87-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...