2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC5335-100G PC5335-25G PC5335-5G
77416-680EA 421.78 USD
77416-680 77416-682 77416-684
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98%
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol
Formula: C₆H₆F₈O₂
Boiling Pt: 100 °C/3 mmHg
Melting Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00069087
CAS Number: 355-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR