1H,1H,2H,2H-Heneicosafluoro-1-dodecanol 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC5996-1G PC5996-250MG
77417-242EA 63.16 USD
77417-242 77417-244
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol 97%
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol
Formula: C₁₂H₅F₂₁O
Boiling Pt: 145 °C/10 mmHg
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00039545
CAS Number: 865-86-1
EINECS: 212-748-7
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR