Propagyl bromide 80% in toluene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Bromopropyne

OR01606-250ML OR01606-50ML
77401-778EA 412.62 USD
77401-778 77401-780
Propagyl bromide 80% in toluene
Propagyl bromide
Formula: C₃H₃Br
MW: 118.96 g/mol
Boiling Pt: 90
Melting Pt: -43
Density: 1.335 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 2345
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR