[2.2]-Paracyclophane 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Tricyclo[8,2,2,24,7]hexadeca-4,6,10,12,13,15-hexaene, Tricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene
OR18296-100G
OR18296-25G
OR18296-5G
77406-208EA
115.12
USD
77406-208
77406-210
77406-212
[2.2]-Paracyclophane 98%
[2.2]-Paracyclophane
Formula:
C₁₆H₁₆ MW: 208.3 g/mol Melting Pt: 283…288 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003707 CAS Number: 1633-22-3 EINECS: 216-644-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...