4-Fluoro-2-methylbenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Fluoro-2-methylbenzene-1-sulphonyl chloride

PC6571-25G PC6571-5G
77417-444EA 395.44 USD
77417-444 77417-446
4-Fluoro-2-methylbenzenesulfonyl chloride
4-Fluoro-2-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Boiling Pt: 245…246 °C
Density: 1.433 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094390
CAS Number: 7079-48-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR