Ethyl heptafluorobutyrate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethyl 2,2,3,3,4,4,4-heptafluorobutanoate, Ethyl perfluorobutyrate

PC3240-100G PC3240-25G PC3240-5G
77414-592EA 192.56 USD
77414-592 77414-594 77414-596
Ethyl heptafluorobutyrate 98%
Ethyl heptafluorobutyrate
Formula: C₆H₅F₇O₂
MW: 242.09 g/mol
Boiling Pt: 95
Density: 1.396 g/cm³ (25 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00000434
CAS Number: 356-27-4
EINECS: 206-602-1
UN: 3272
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR