3-Fluoro-4-hydroxybenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC9903-25G PC9903-5G
77418-758EA 44.4 USD
77418-758 77418-760
3-Fluoro-4-hydroxybenzonitrile
3-Fluoro-4-hydroxybenzonitrile
Formula: C₇H₄FNO
MW: 137.11
Melting Pt: 132…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00215834
CAS Number: 405-04-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR