Bis(4-nitrophenyl)carbonate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR16983-100G OR16983-25G OR16983-500G
77405-868EA 47.88 USD
77405-868 77405-870 77405-872
Bis(4-nitrophenyl)carbonate 98%
Bis(4-nitrophenyl)carbonate
Formula: C₁₃H₈N₂O₇
Melting Pt: 136…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007322
CAS Number: 5070-13-3
EINECS: 225-775-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR