(1S)-(+)-Camphor-10-sulfonic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR922840-100G OR922840-500G
77411-822EA 54 USD
77411-822 77411-824
(1S)-(+)-Camphor-10-sulfonic acid 98%
(1S)-(+)-Camphor-10-sulphonic acid
Formula: C₁₀H₁₆O₄S
Melting Pt: 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00064157
CAS Number: 3144-16-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR