2,4-Dichloro-6-methylbenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,4-Dichloro-6-methylbenzene-1-sulphonyl chloride

OR30237-1G OR30237-500MG OR30237-5G
77408-832EA 108 USD
77408-832 77408-834 77408-836
2,4-Dichloro-6-methylbenzenesulfonyl chloride
2,4-Dichloro-6-methylbenzenesulphonyl chloride
Formula: C₇H₅Cl₃O₂S
MW: 259.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00204179
CAS Number: 175278-26-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR