α'-Chloro-α,α,α-trifluoro-o-xylene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7633E-100G PC7633E-25G PC7633E-5G
77418-054EA 110.04 USD
77418-054 77418-056 77418-058
α'-Chloro-α,α,α-trifluoro-o-xylene 97%
α'-Chloro-α,α,α-trifluoro-o-xylene
Formula: C₈H₆ClF₃
Boiling Pt: 67…68 °C/12 mmHg
Density: 1.337 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00009920
CAS Number: 21742-00-7
EINECS: 244-560-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR