4'-Chloro-3'-nitroacetophenone
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Chloro-3-nitroacetophenone
OR0027-25G
OR0027-5G
77401-638EA
44.82
USD
77401-638
77401-640
4'-Chloro-3'-nitroacetophenone
4'-Chloro-3'-nitroacetophenone
Formula:
C₈H₆ClNO₃ MW: 199.59 g/mol Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007083 CAS Number: 5465-65-6 EINECS: 226-769-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...