2-Bromo-4,6-difluorobenzenesulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC9618-1G PC9618-5G
77418-698EA 21.26 USD
77418-698 77418-700
2-Bromo-4,6-difluorobenzenesulfonyl chloride 95%
2-Bromo-4,6-difluorobenzenesulphonyl chloride
Formula: C₆H₂BrClF₂O₂S
MW: 291.5 g/mol
Boiling Pt: 265…267 °C
Density: 1.93 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094561
CAS Number: 351003-42-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR