4-Bromo-2-fluoro-5-iodotoluene 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC8831-1G
PC8831-25G
PC8831-5G
77418-456EA
21.26
USD
77418-456
77418-458
77418-460
4-Bromo-2-fluoro-5-iodotoluene 95%
4-Bromo-2-fluoro-5-iodotoluene
Formula:
C₇H₅BrFI Boiling Pt: 249…250 °C Density: 2.145 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD06200719 CAS Number: 870704-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...