4-Bromo-3-fluorobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC9579-100G PC9579-25G PC9579-5G
77418-680EA 227.2 USD
77418-680 77418-682 77418-684
4-Bromo-3-fluorobenzonitrile 95%
4-Bromo-3-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Melting Pt: 92…93 °C
Storage Temperature: Ambient
MDL Number: MFCD07782069
CAS Number: 133059-44-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR