2-Bromo-6-fluorobenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7441-100G PC7441-25G PC7441-5G
77417-984EA 227.2 USD
77417-984 77417-986 77417-988
2-Bromo-6-fluorobenzaldehyde 97%
2-Bromo-6-fluorobenzaldehyde
Formula: C₇H₄BrFO
Boiling Pt: 100…102 °C/8 mmHg
Melting Pt: 39…47 °C
Density: 1.7 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD03407341
CAS Number: 360575-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR