5-Bromo-2-chloro-4-methyl-3-nitropyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR2915-10G OR2915-1G OR2915-5G
77408-346EA 70.3 USD
77408-346 77408-348 77408-350
5-Bromo-2-chloro-4-methyl-3-nitropyridine 95%
5-Bromo-2-chloro-4-methyl-3-nitropyridine
Formula: C₆H₄BrClN₂O₂
MW: 251.47
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD06659505
CAS Number: 884495-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR